Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

223522

Sigma-Aldrich

Sodium phosphate monobasic monohydrate

≥98%, ACS reagent

Synonym(s):

Monosodium phosphate, Sodium dihydrogen phosphate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaH2PO4 · H2O
CAS Number:
Molecular Weight:
137.99
EC Number:
MDL number:
UNSPSC Code:
12161700
Pricing and availability is not currently available.

grade

ACS reagent

Assay

≥98%
98.0-102.0%

impurities

≤0.01% Ca ppt.
≤0.01% NH4OH pptd.
≤0.01% insolubles pptd.

pH

4.1-4.5

anion traces

chloride (Cl-): ≤5 ppm
sulfate (SO42-): ≤0.003%

cation traces

Fe: ≤0.001%
heavy metals: ≤0.001%

SMILES string

[Na+].[H]O[H].OP(O)([O-])=O

InChI

1S/Na.H3O4P.H2O/c;1-5(2,3)4;/h;(H3,1,2,3,4);1H2/q+1;;/p-1

InChI key

BBMHARZCALWXSL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in poly bottles

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service