Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

223212

Sigma-Aldrich

Tetramethylammonium hydroxide pentahydrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(OH) · 5H2O
CAS Number:
Molecular Weight:
181.23
Beilstein:
3714235
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

1.1 (vs air)

Assay

97%

autoignition temp.

878 °F

expl. lim.

36 %

mp

67-70 °C (lit.)

SMILES string

O.O.O.O.O.[OH-].C[N+](C)(C)C

InChI

1S/C4H12N.6H2O/c1-5(2,3)4;;;;;;/h1-4H3;6*1H2/q+1;;;;;;/p-1

InChI key

MYXKPFMQWULLOH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 1 Dermal - STOT SE 1

Target Organs

Central nervous system, Liver,thymus

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Shen et al.
Chemical communications (Cambridge, England), 49(73), 8114-8116 (2013-08-08)
A simple and low-cost colorimetric assay utilizing ferrofluidic nanoparticulate probes (FNPs) and a ligase for single-nucleotide polymorphism genotyping is described. Excellent sensitivity and selectivity were accomplished through the engagement of the FNPs and a ligase chain reaction.
Marco I Ries et al.
The Journal of biological chemistry, 288(52), 37289-37295 (2013-11-15)
Metabolic profiling and structural elucidation of novel secondary metabolites obtained from derived deletion strains of the filamentous fungus Penicillium chrysogenum were used to reassign various previously ascribed synthetase genes of the roquefortine/meleagrin pathway to their corresponding products. Next to the
Jacob O Fierer et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(13), E1176-E1181 (2014-03-19)
Individual proteins can now often be modified with atomic precision, but there are still major obstacles to connecting proteins into larger assemblies. To direct protein assembly, ideally, peptide tags would be used, providing the minimal perturbation to protein function. However
Markus Welcker et al.
Genes & development, 27(23), 2531-2536 (2013-12-04)
The Fbw7 tumor suppressor targets a broad network of proteins for ubiquitylation. Here we show critical functions for Fbw7 dimerization in regulating the specificity and robustness of degradation. Dimerization enables Fbw7 to target substrates through concerted binding to two suboptimal
Mei Fang et al.
Nanotechnology, 23(14), 145601-145601 (2012-03-22)
Magnetite nanoparticles have been prepared by co-precipitation using a custom-designed jet mixer to achieve rapid mixing (RM) of reactants in a timescale of milliseconds. The quick and stable nucleation obtained allows control of the particle size and size distribution via

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service