Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

222828

Sigma-Aldrich

4-Methoxy-3-nitrobenzotrifluoride

99%

Synonym(s):

3-Nitro-4-methoxy-α,α,α-trifluorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(NO2)CF3
CAS Number:
Molecular Weight:
221.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

bp

152-153 °C/28 mmHg (lit.)

mp

47-49 °C (lit.)

SMILES string

COc1ccc(cc1[N+]([O-])=O)C(F)(F)F

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

8-Hydroxy-5-trifluoromethylquinoline.
Pettit MR and Tatlow JC.
Journal of the Chemical Society, 3852-3854 (1954)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service