Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

222798

Sigma-Aldrich

N-Methylbenzamide

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CONHCH3
CAS Number:
Molecular Weight:
135.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

solid

bp

167 °C/11 mmHg (lit.)

mp

76-78 °C (lit.)

solubility

ethanol: soluble 50 mg/mL, clear, yellow-green

functional group

amide
phenyl

SMILES string

CNC(=O)c1ccccc1

InChI

1S/C8H9NO/c1-9-8(10)7-5-3-2-4-6-7/h2-6H,1H3,(H,9,10)

InChI key

NCCHARWOCKOHIH-UHFFFAOYSA-N

General description

N-Methylbenzamide is a potent PDE10A (phosphodiesterase with a remarkable localization as the protein is abundant only in brain tissue) inhibitor[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H van de Waterbeemd et al.
Journal of medicinal chemistry, 29(5), 600-606 (1986-05-01)
The electronic properties of orthopramides, a group of selective D-2 dopamine receptor antagonists, were investigated by calculating molecular electrostatic potentials (MEP) of model compounds with the ab initio STO-3G MO method. The various substitution patterns of the aromatic ring are
L Constantino et al.
Biochemical pharmacology, 44(4), 651-658 (1992-08-18)
The metabolism of N,N-dimethylbenzamides by phenobarbital-induced rat liver microsomes results in the formation of N-methylbenzamides and formaldehyde. The reaction proceeds via the formation of an intermediate N-hydroxymethyl-N-methylbenzamide, which, for the microsomal oxidation of N,N-dimethylbenzamide, was isolated and characterized. Confirmation of
Atsushi Satoh et al.
Bioorganic & medicinal chemistry letters, 19(18), 5464-5468 (2009-08-14)
We identified 4-fluoro-N-[4-[6-(isopropylamino)pyrimidin-4-yl]-1,3-thiazol-2-yl]-N-methylbenzamide 27 as a potent mGluR1 antagonist. The compound possessed excellent subtype selectivity and good PK profile in rats. It also demonstrated relatively potent antipsychotic-like effects in several animal models. Suitable for development as a PET tracer, compound
John Paul Kilburn et al.
Bioorganic & medicinal chemistry, 21(19), 6053-6062 (2013-08-28)
PDE10A is a recently identified phosphodiesterase with a quite remarkable localization since the protein is abundant only in brain tissue. Based on this unique localization, research has focused extensively on using PDE10A modulators as a novel therapeutic approach for dysfunction
Shih-Chung Huang et al.
Bioorganic & medicinal chemistry letters, 14(18), 4779-4782 (2004-08-25)
A series of N-methylbenzamide analogues (2-18) that is structurally derived from SR 48,968, a potent neurokinin-2 (NK(2)) receptor antagonist (pK(b)9.1), has been obtained using asymmetric synthesis. Isothiocyanato-N-methylbenzamide (10-12) and bromoacetamido-N-methylbenzamide derivatives (16-18) have been designed to serve as potential electrophilic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service