Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

222275

Sigma-Aldrich

2-Thiopheneglyoxylic acid

95%

Synonym(s):

α-Oxo-2-thiopheneacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$137.00

$137.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$137.00

About This Item

Empirical Formula (Hill Notation):
C6H4O3S
CAS Number:
Molecular Weight:
156.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$137.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

95%

mp

88-91 °C (lit.)

SMILES string

OC(=O)C(=O)c1cccs1

InChI

1S/C6H4O3S/c7-5(6(8)9)4-2-1-3-10-4/h1-3H,(H,8,9)

InChI key

GIWRVUADKUVEGU-UHFFFAOYSA-N

Application

2-Thiopheneglyoxylic acid was used in the synthesis of europium complexes[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Yu et al.
Guang pu xue yu guang pu fen xi = Guang pu, 24(12), 1571-1574 (2005-04-15)
The complex of europium with 1,10-phenanthroline and 2-thiopheneglyoxylic acid (HL) was synthesized. Its chemical formula was determined to be [EuL2 x phen x (H2O)3]NO3 by elemental analysis, electrical conductivity, IR and 1H NMR. It was found that the europium ion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service