Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

221902

Sigma-Aldrich

(1R,2R,3R,5S)-(−)-Isopinocampheol

98%

Synonym(s):

(−)-Isopinocampheol, (1R,2R,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptan-3-ol, (1R,2R,3R,5S)-3-Pinanol, 3-Pinanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
2038100
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

optical activity

[α]22/D −34°, c = 20 in ethanol

optical purity

ee: 95% (GLC)

bp

219 °C (lit.)

mp

51-53 °C (lit.)

functional group

hydroxyl

SMILES string

C[C@H]1[C@H](O)CC2CC1C2(C)C

InChI

1S/C10H18O/c1-6-8-4-7(5-9(6)11)10(8,2)3/h6-9,11H,4-5H2,1-3H3/t6-,7+,8-,9-/m1/s1

InChI key

REPVLJRCJUVQFA-BZNPZCIMSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

200.1 °F - closed cup

Flash Point(C)

93.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service