Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

220337

Sigma-Aldrich

7-Methoxycoumarin

≥98%

Synonym(s):

Herniarin, Methyl umbelliferyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O3
CAS Number:
Molecular Weight:
176.17
Beilstein:
141728
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98%

mp

117-121 °C (lit.)

SMILES string

COc1ccc2C=CC(=O)Oc2c1

InChI

1S/C10H8O3/c1-12-8-4-2-7-3-5-10(11)13-9(7)6-8/h2-6H,1H3

InChI key

LIIALPBMIOVAHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Legrum et al.
The Journal of pharmacology and experimental therapeutics, 221(3), 790-794 (1982-06-01)
Pretreatment of rats with cobaltous chloride has been shown previously to reduce the content of cytochrome P-450 in the hepatic microsomal protein. This is accompanied by a corresponding decrease in substrate oxidation, e.g. ethyl morphine demethylation, in vitro. The present
Prince Firdoos Iqbal et al.
European journal of medicinal chemistry, 44(5), 2252-2259 (2008-08-05)
In continuation of our search for potential antiamoebic agents from folklore Indian medicinal plants, we found that the benzene and ethyl acetate extracts from the root bark of Adina cordifolia exhibited strong antiamoebic activity with IC(50) values of 2.92 and
Chao-Mei Ma et al.
Phytochemical analysis : PCA, 18(1), 42-49 (2007-01-31)
A simple HPLC-PAD-MS method was established to quantitatively analyse two spiroether isomers (cis-en-yn-dicycloether and trans-en-yn-dicycloether) and the main coumarin, herniarin, in chamomile herbs, simultaneously. By using this method, the contents of these three compounds in the flowers of two chamomile
P Paterson et al.
Xenobiotica; the fate of foreign compounds in biological systems, 14(11), 849-859 (1984-11-01)
Pretreatment of rats with phenobarbitone increased hepatic microsomal 7-methoxy-and 7-ethoxy-coumarin O-dealkylase activities. Pretreatment with beta-naphthoflavone increased only the 7-ethoxycoumarin O-dealkylase activity. The addition of metyrapone in vitro inhibited the O-dealkylations to different extents. Similar results were obtained with diphenyloxazole and
Adriana Eliasová et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 59(7-8), 543-548 (2005-04-09)
The responses of young plants of diploid and tetraploid Matricaria chamomilla cultivars to abiotic stress were studied. The course of quantitative changes of main leaf secondary metabolites was evaluated within an interval from 6 h before to 54 h after

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service