Skip to Content
MilliporeSigma
All Photos(1)

Documents

219401

Sigma-Aldrich

2-Fluoroiodobenzene

99%, contains copper as stabilizer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4I
CAS Number:
Molecular Weight:
222.00
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.59 (lit.)

bp

188-189 °C (lit.)

mp

−41-−40 °C (lit.)

density

1.903 g/mL at 25 °C (lit.)

SMILES string

Fc1ccccc1I

InChI

1S/C6H4FI/c7-5-3-1-2-4-6(5)8/h1-4H

InChI key

TYHUGKGZNOULKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Fluoroiodobenzene was used in the synthesis of:
  • bridged biphenyl compounds, 1-fluoro-8-methylbiphenylene and 4-fluoro-5-methylfluorene
  • 2-acetyl-7-iodo-1-benzothiophene
  • chiral o-phosphanophenyl sulfoxides

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierre-Yves Michellys et al.
Bioorganic & medicinal chemistry letters, 14(6), 1593-1598 (2004-03-10)
The synthesis and in vitro characterization of novel RXR-selective ligands possessing various substituted 1-benzofuran or 1-benzothiophene moieties are described.
Long-range proton-fluorine spin-spin coupling in bridged biphenyls. Compelling evidence for a" through-space"(" direct") mechanism.
Gribble GW and Douglas Jr JR.
Journal of the American Chemical Society, 92(19), 5764-5765 (1970)
New chiral sulfoxide ligands possessing a phosphano or phosphanoamino functionality in palladium-catalyzed asymmetric allylic nucleophilic substitution reactions.
Hiroi K, et al.
Tetrahedron, 56(27), 4701-4710 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service