Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

219363

Sigma-Aldrich

2-Fluorobenzotrifluoride

99%

Synonym(s):

α,α,α,Ar-Tetrafluorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CF3
CAS Number:
Molecular Weight:
164.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

liquid

refractive index

n20/D 1.406 (lit.)

bp

114-115 °C/750 mmHg (lit.)

density

1.293 g/mL at 25 °C (lit.)

SMILES string

Fc1ccccc1C(F)(F)F

InChI

1S/C7H4F4/c8-6-4-2-1-3-5(6)7(9,10)11/h1-4H

InChI key

BGVGHYOIWIALFF-UHFFFAOYSA-N

Application

2-Fluorobenzotrifluoride was used in the synthesis of 2,2′-bis(trifluoromethylphenoxy)biphenyl via nucleophilic aromatic substitution reaction with 2,2′-biphenol.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akiko Okamoto et al.
Journal of oleo science, 56(9), 479-491 (2007-09-28)
Novel bifunctional acyl-acceptant biphenyls bearing trifluoromethylated aroyloxy groups were successfully synthesized in high yields via TfOH-mediated electrophilic aromatic aroylation of fluorobenzene with CF(3)-bearing aroyl chlorides followed by nucleophilic aromatic substitution with 2,2'-biphenol. Similarly, 2,2'-bis(trifluoromethylphenoxy)biphenyl was synthesized via nucleophilic aromatic substitution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service