Skip to Content
MilliporeSigma
All Photos(1)

Documents

219231

Sigma-Aldrich

N,N-Dimethyl-p-phenylenediamine dihydrochloride

99%

Synonym(s):

4-Amino-N,N-dimethylaniline dihydrochloride, DMPD · 2HCl, DMPPDA · 2HCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4NH2 · 2HCl
CAS Number:
Molecular Weight:
209.12
Beilstein:
3913463
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

99%

mp

222 °C (dec.) (lit.)

SMILES string

Cl[H].Cl[H].CN(C)c1ccc(N)cc1

InChI

1S/C8H12N2.2ClH/c1-10(2)8-5-3-7(9)4-6-8;;/h3-6H,9H2,1-2H3;2*1H

InChI key

IAEDWDXMFDKWFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F J Alcaín et al.
Redox report : communications in free radical research, 3(5-6), 287-293 (1998-12-16)
Swiss 3T3 fibroblasts can be weakly stimulated to grow by bombesin, epidermal growth factor or ceruloplasmin when cells are maintained in Dulbecco's Modified Essential Medium (DMEM), the pH of which is 7.75. Addition of insulin synergizes with the other mitogens.
P K Wong et al.
Journal of applied microbiology, 85(1), 79-87 (1998-08-29)
Klebsiella pneumoniae RS-13 and Acetobacter liquefaciens S-1, both methyl red (MR)-degrading bacterial strains, degraded N,N'-dimethyl-p-phenylenediamine (DMPD) under aerobic conditions. DMPD, a toxic and mutagenic aromatic amine, is formed during the reductive cleavage of azo dyes such as MR. The effects
Claudineia R Silva et al.
Talanta, 85(3), 1703-1705 (2011-08-03)
A spectrophotometric flow injection procedure involving N,N-dimethyl-p-phenylenediamine (DMPD) is applied to the sulfide monitoring of a sugar fermentation by Saccharomyces cerevisiae under laboratory conditions. The gaseous chemical species evolving from the fermentative process, mainly CO(2), are trapped allowing a cleaned
C Li et al.
The Biochemical journal, 300 ( Pt 1), 31-36 (1994-05-15)
C-terminal alpha-amidation is a structural feature essential to the biological activity of many peptide hormones. Peptidylglycine alpha-amidating mono-oxygenase (PAM; EC 1.14.17.3) catalyses conversion of glycine-extended peptide hormone precursors into their corresponding alpha-hydroxyglycine derivatives. This reaction is the first step in
C Störle et al.
Chemico-biological interactions, 83(3), 271-291 (1992-08-28)
N,N-Dimethyl-p-phenylenediamine (DMPD) reacted directly with oxyhemoglobin under formation of ferrihemoglobin and, presumably, the N,N-dimethyl-p-phenylenediamine radical cation (DMPP.+). The apparent second-order rate constant of this reaction was 1 M-1 s-1 (pH 7.4, 37 degrees C). The reaction rate was diminished by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service