Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

219177

Sigma-Aldrich

4,5-Dimethylthiazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NS
CAS Number:
Molecular Weight:
113.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

refractive index

n20/D 1.521 (lit.)

bp

158 °C/742 mmHg (lit.)

density

1.07 g/mL at 25 °C (lit.)

SMILES string

Cc1ncsc1C

InChI

1S/C5H7NS/c1-4-5(2)7-3-6-4/h3H,1-2H3

InChI key

UWSONZCNXUSTKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Radical bromination of 4,5-dimethylthiazole using N-bromosuccinimide in the presence of 2,2′-azobisisobutyronitrile yields mono-, tri- and tetrabromo compounds.

Application

4,5-Dimethylthiazole was used in the preparation of 4-(bromomethyl)-5-(dibromomethyl)thiazole.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mouaffak Al Hariri et al.
The Journal of organic chemistry, 62(2), 405-410 (1997-01-24)
4-(Bromomethyl)-5-(dibromomethyl)thiazole (1) was prepared in good yields by bromination of 4,5-dimethylthiazole with 3.3 equiv of NBS in the presence of AIBN. Treatment of 1 with sodium iodide led to a thiazole o-quinodimethane 2 which was trapped in situ with dienophiles
Selective Bromination of 4, 5-Dimethylthiazole with N-Bromosuccinimide.
Al Hariri M, et al.
European Journal of Organic Chemistry, 1998(4), 593-594 (1998)
Le Wu et al.
Journal of molecular neuroscience : MN, 56(4), 848-857 (2015-02-24)
Lipoxin A4 (LXA4), a potent antioxidant and anti-inflammation mediator, protects brains against cerebral ischemia/reperfusion (I/R) injury in vivo. However, few reports concern its function on astrocytes during cerebral I/R injury. The pathogenesis of cerebral I/R injury involves oxidative stress caused
João G Marques et al.
Pharmaceutical research, 31(9), 2516-2528 (2014-03-14)
Cancer multi-drug resistance is a major issue associated with current anti-tumoral therapeutics. In this work, Crizotinib an anti-tumoral drug approved for the treatment of non-small lung cancer in humans, and Sildenafil (Viagra(®)), were loaded into micellar carriers to evaluate the
Szu-Jung Chen et al.
Inflammation, 38(2), 534-545 (2014-07-06)
Eicosatrienoic acid (Δ11,14,17-20:3; ETrA) is a rare naturally occurring n-3 polyunsaturated fatty acid (PUFA). Using murine RAW264.7 cells, the objectives were to determine how ETrA modulated phospholipid fatty acid compositions and the production of pro-inflammatory mediators. Incubation cells with ETrA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service