Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

218448

Sigma-Aldrich

4-Nitroquinoline N-oxide

98%

Synonym(s):

4-NQO, 4-Nitroquinoline 1-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6N2O3
CAS Number:
Molecular Weight:
190.16
Beilstein:
165756
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

98%

mp

154-156 °C (lit.)

SMILES string

[O-][N+](=O)c1cc[n+]([O-])c2ccccc12

InChI

1S/C9H6N2O3/c12-10-6-5-9(11(13)14)7-3-1-2-4-8(7)10/h1-6H

InChI key

YHQDZJICGQWFHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leon F Stankowski et al.
Environmental and molecular mutagenesis, 52(9), 738-747 (2011-10-25)
As part of the Stage III Pig-a multilaboratory validation trial, we examined the induction of CD59-negative reticulocytes and total red blood cells (RET(CD59-) and RBC(CD59-) , respectively) in male Sprague Dawley(®) rats treated with 4-nitroquinoline-1-oxide (4NQO), for 28 consecutive days
Francesca Trotta et al.
International journal of food microbiology, 153(3), 275-280 (2011-12-20)
Yeasts isolated from Italian beverages and foods (wine and cheeses) were identified as Saccharomyces cerevisiae and Debaryomyces hansenii by sequencing the D1/D2 domain of the 26S rRNA gene and differentiated, at strain level, by microsatellite PCR fingerprinting and RAPD-PCR. All
Yu-Ching Liu et al.
Toxicology and applied pharmacology, 262(2), 107-116 (2012-05-09)
The purpose of this study was to identify the genes induced early in murine oral carcinogenesis. Murine tongue tumors induced by the carcinogen, 4-nitroquinoline 1-oxide (4-NQO), and paired non-tumor tissues were subjected to microarray analysis. Hierarchical clustering of upregulated genes
J Peter Svensson et al.
PloS one, 7(5), e37368-e37368 (2012-05-23)
Studies in Saccharomyces cerevisiae show that many proteins influence cellular survival upon exposure to DNA damaging agents. We hypothesized that human orthologs of these S. cerevisiae proteins would also be required for cellular survival after treatment with DNA damaging agents.
Kwame Osei-Sarfo et al.
Carcinogenesis, 34(11), 2673-2681 (2013-06-21)
Head and neck squamous cell carcinoma (HNSCC) is the sixth most common type of cancer affecting humans worldwide. To determine the potential mechanisms by which chronic tobacco and alcohol abuse lead to HNSCC of the oral cavity, we have used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service