Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

217204

Sigma-Aldrich

Neodymium(III) nitrate hydrate

99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Nd(NO3)3 · xH2O
CAS Number:
Molecular Weight:
330.25 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99.99% trace metals basis

form

crystalline

composition

Degree of hydration, 5-6

reaction suitability

reagent type: catalyst
core: neodymium

impurities

≤200.0 ppm Trace Rare Earth Analysis

SMILES string

O.[Nd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/3NO3.Nd.H2O/c3*2-1(3)4;;/h;;;;1H2/q3*-1;+3;

InChI key

GZIAUVGSMHERLN-UHFFFAOYSA-N

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel González-Mancebo et al.
Journal of colloid and interface science, 520, 134-144 (2018-03-14)
The singular properties of lanthanide-based inorganic nanoparticles (NPs) has raised the attention of the scientific community in biotechnological applications. In particular, those systems with two or more functionalities are especially interesting. In this work, an effective and commercially attractive procedure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service