Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

215570

Sigma-Aldrich

1,4-Cyclohexanedione mono(2,2-dimethyltrimethylene ketal)

95%

Synonym(s):

3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18O3
CAS Number:
Molecular Weight:
198.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

form

solid

mp

45-50 °C (lit.)

SMILES string

CC1(C)COC2(CCC(=O)CC2)OC1

InChI

1S/C11H18O3/c1-10(2)7-13-11(14-8-10)5-3-9(12)4-6-11/h3-8H2,1-2H3

InChI key

COKVDTKAWIFNTH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,4-Cyclohexanedione mono(2,2-dimethyltrimethylene ketal) has been used in the synthesis of:
  • 3,4-dihydro-2(1H)-naphthalenones via carbainion induced reaction[1]
  • 8-aryl-5-cyano-6-sec-amino-3,4-dihydro-2(1H)-naphthalenone-(2,2-dimethyltrimethylene)ketals[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V J Ram et al.
The Journal of organic chemistry, 66(16), 5333-5337 (2001-08-04)
An innovative approach to the one-pot synthesis of highly functionalized 3,4-dihydro-1H-isothiochromenes (3), 6H-benzo[c]thiochromenes (5, 6), 6H-benzo[c]chromenes (8), and 2,3-dihydro-1-benzothiophenes (10, 11) is delineated from the reaction of a suitably functionalized 6-aryl-3-carbomethoxy-4-methylthio-2H-pyran-2-one (1) and a carbanion generated from tetrahydrothiopyran-4-one, 4-thiochromanone, 4-chromanone

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service