Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

215341

Sigma-Aldrich

3,5-Dibenzyloxyacetophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2O)2C6H3COCH3
CAS Number:
Molecular Weight:
332.39
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

60-62 °C (lit.)

functional group

ketone
phenyl

SMILES string

CC(=O)c1cc(OCc2ccccc2)cc(OCc3ccccc3)c1

InChI

1S/C22H20O3/c1-17(23)20-12-21(24-15-18-8-4-2-5-9-18)14-22(13-20)25-16-19-10-6-3-7-11-19/h2-14H,15-16H2,1H3

InChI key

KOJXGMJOTRYLBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,5-Dibenzyloxyacetophenone was used in the synthesis of enantiomerically pure B-ring modified analogs of (−)-epicatechin gallate[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James C Anderson et al.
Tetrahedron, 61(32), 7703-7711 (2005-08-08)
Two enantiomerically pure B-ring modified analogues of (-)-epicatechin gallate were synthesised and their modulation of beta-lactam resistance using three strains of methicillin resistant Staphylococcus aureus (BB 568, EMRSA-15 and EMRSA-16) evaluated. Sub-inhibitory concentrations (12.5 and 25 mg/L) of the two

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service