Skip to Content
MilliporeSigma
All Photos(1)

Documents

215295

Sigma-Aldrich

cis-2-Methylcyclohexanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H10OH
CAS Number:
Molecular Weight:
114.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.465 (lit.)

bp

165 °C (lit.)

mp

6-8 °C (lit.)

density

0.936 g/mL at 25 °C (lit.)

SMILES string

C[C@@H]1CCCC[C@@H]1O

InChI

1S/C7H14O/c1-6-4-2-3-5-7(6)8/h6-8H,2-5H2,1H3/t6-,7+/m1/s1

InChI key

NDVWOBYBJYUSMF-RQJHMYQMSA-N

General description

Elimination reaction of cis-2-methylcyclohexanol catalyzed by alumina and phosphoric acid has been studied. Dehydration of cis-2-methylcyclohexanol over basic catalysts (alkaline earth oxides) has been reported.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elimination Reactions of 2-Methylcyclohexyl Derivatives.
Botteron DG and Shulman GP.
The Journal of Organic Chemistry, 27(6), 2007-2010 (1962)
Catalytic dehydration of secondary alcohols over thoria: Infrared study and generalization to basic catalysts.
Canesson P and Blanchard M.
J. Catal., 42(2), 205-212 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service