Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

214728

Sigma-Aldrich

Boron

crystalline, 90-95%, −325 mesh

Synonym(s):

Boron atom, Boron element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
B
CAS Number:
Molecular Weight:
10.81
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

Assay

90-95%

form

crystalline

reaction suitability

reagent type: catalyst
core: boron

resistivity

1.5E12 μΩ-cm, 20 °C

particle size

−325 mesh

density

2.34 g/mL at 25 °C (lit.)

SMILES string

[B]

InChI

1S/B

InChI key

ZOXJGFHDIHLPTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan Fietzke et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(10), 2960-2965 (2015-02-26)
No records exist to evaluate long-term pH dynamics in high-latitude oceans, which have the greatest probability of rapid acidification from anthropogenic CO2 emissions. We reconstructed both seasonal variability and anthropogenic change in seawater pH and temperature by using laser ablation
Wenyao Chen et al.
ChemSusChem, 8(17), 2927-2931 (2015-06-11)
We report remarkable support effects for carbon nanotubes (CNTs) in the Pt/CNT-catalyzed hydrolytic dehydrogenation of ammonia borane. The origin of the support-dependent activity and durability is elucidated by combining the catalytic and durability testing with characterization by a range of
Linda K Månsson et al.
Faraday discussions, 181, 49-69 (2015-05-15)
We describe a route towards self-assembled colloidal molecules, where thermoresponsive microgels serve as discrete, externally tunable interaction sites. The ability of poly(N-isopropylacrylamide) (PNIPAM) and poly(N-isopropylmethacrylamide) (PNIPMAM) microgels to adsorb to the oil/water (O/W) interface and create Pickering-stabilized mini-emulsions was first
Wenwen Xu et al.
Soft matter, 11(36), 7151-7158 (2015-08-11)
Anisotropic particles have generated an enormous amount of research interest due to their applications for drug delivery, electronic displays and as micromotors. However, up till now, there is no single protocol capable of generating particles of "patchy" composition with a
Syeda Shegufta Ameer et al.
Environmental research, 140, 32-36 (2015-04-01)
Exposure to inorganic arsenic has been identified as a risk factor for elevated blood pressure and cardiovascular disease. Our aim with this study was to elucidate effects of arsenic on blood pressure and early risk markers of cardiovascular disease in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service