Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

214493

Sigma-Aldrich

Chrysoidine G

Dye content 90 %

Synonym(s):

4-Phenylazo-m-phenylenediamine monohydrochloride, Basic Orange 2, Chrysoidine Y

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N4 · HCl
CAS Number:
Molecular Weight:
248.71
Colour Index Number:
11270
Beilstein:
3724653
EC Number:
MDL number:
UNSPSC Code:
12171500

composition

Dye content, 90%

mp

235 °C (dec.) (lit.)

λmax

449 nm

SMILES string

Cl[H].Nc1ccc(N=Nc2ccccc2)c(N)c1

InChI

1S/C12H12N4.ClH/c13-9-6-7-12(11(14)8-9)16-15-10-4-2-1-3-5-10;/h1-8H,13-14H2;1H

InChI key

MCTQNEBFZMBRSQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Muta. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[A bile-chrysoidine-glycerol culture medium and its use in the diagnosis of gram-negative aerobic bacteria, especially Enterobacteriaceae].
K Ziesché et al.
Zeitschrift fur die gesamte Hygiene und ihre Grenzgebiete, 31(9), 516-518 (1985-09-01)
Rhiannon M David et al.
Environmental toxicology and chemistry, 28(9), 1893-1900 (2009-04-24)
The use of unicellular algae in ecotoxicity testing is well established, particularly regarding whole-organism and population-level end points such as lethality and population growth. Conflicting information exists, however, on the potential for genetic toxicity to be incorporated into the safety
Maggots dyed with chrysoidine.
British medical journal (Clinical research ed.), 289(6456), 1451-1452 (1984-11-24)
[Hobby and occupation as causes of cancer].
R L Zielhuis
Nederlands tijdschrift voor geneeskunde, 130(22), 994-998 (1986-05-31)
Maggots dyed with chrysoidine: a possible risk to anglers.
G M Sole
British medical journal (Clinical research ed.), 289(6451), 1043-1044 (1984-10-20)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service