Skip to Content
MilliporeSigma
All Photos(1)

Documents

21335

Sigma-Aldrich

(1R)-Camphor oxime

≥97.0% (sum of enantiomers, GC)

Synonym(s):

(1R)-1,7,7-Trimethylbicylco[2,2,1]heptan-2-one oxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO
CAS Number:
Molecular Weight:
167.25
Beilstein:
2413664
EC Number:
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:

Quality Level

Assay

≥97.0% (sum of enantiomers, GC)

form

solid

optical activity

[α]20/D −40.5±1°, c = 5% in ethanol

mp

115-119 °C

SMILES string

CC1(C)[C@@H]2CC[C@@]1(C)C(\C2)=N\O

InChI

1S/C10H17NO/c1-9(2)7-4-5-10(9,3)8(6-7)11-12/h7,12H,4-6H2,1-3H3/b11-8+/t7-,10+/m1/s1

InChI key

OVFDEGGJFJECAT-RZIOALPKSA-N

Other Notes

Chiral intermediate; starting material for the preparation of R(+)-bornylamine and R(-)isobornylamine

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W. Huckel et al.
Justus Liebigs Annalen der Chemie, 625, 1-1 (1959)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service