Skip to Content
MilliporeSigma
All Photos(1)

Documents

211338

Sigma-Aldrich

4-Phenylmorpholine

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO
CAS Number:
Molecular Weight:
163.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.63 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

≥98%

form

solid

bp

165-170 °C/45 mmHg (lit.)

mp

51-54 °C (lit.)

SMILES string

C1CN(CCO1)c2ccccc2

InChI

1S/C10H13NO/c1-2-4-10(5-3-1)11-6-8-12-9-7-11/h1-5H,6-9H2

InChI key

FHQRDEDZJIFJAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Phenylmorpholine undergoes oxidation to yield N-formyl-N-2-hydroxyethylaniline.

Application

4-Phenylmorpholine was used as internal standard during the determination of alkaloids by gas chromatographic method.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Different levels of protective alkaloids in grasses with stroma-forming and seed-transmitted Epichloe/Neotyphodium endophytes.
Leuchtmann A, et al.
Journal of Chemical Ecology, 26(4), 1025-1036 (2000)
591. Amine oxidation. Part I. The side-chain oxidation of N-alkyl-and NN-dialkyl-anilines by manganese dioxide.
Henbest HB and Thomas A.
Journal of the Chemical Society, 3032-3039 (1957)
Krishnamoorthy Shanmugaraj et al.
Materials (Basel, Switzerland), 13(4) (2020-02-26)
In this work, Au nanoparticles, supported in Al2O3 nanowires (ANW) modified with (3-aminopropyl)trimethoxysilane were synthetized, for their use as catalysts in the hydrogenation reaction of 4-(2-fluoro-4-nitrophenyl)-morpholine and 4-(4-nitrophenyl)morpholin-3-one. ANW was obtained by hydrothermal techniques and the metal was incorporated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service