Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

210641

Sigma-Aldrich

Acid Red 37

Dye content 65 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17N4NaO8S2
CAS Number:
Molecular Weight:
504.47
Colour Index Number:
17045

Pricing and availability is not currently available. Contact Technical Service

composition

Dye content, 65%


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service