Skip to Content
MilliporeSigma
All Photos(1)

Documents

210269

Sigma-Aldrich

L-allo-Threonine

99%

Synonym(s):

(2S,3S)-2-Amino-3-hydroxybutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH(NH2)CO2H
CAS Number:
Molecular Weight:
119.12
Beilstein:
1721645
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

optical activity

[α]23/D +9.0°, c = 2 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

272 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

C[C@H](O)[C@H](N)C(O)=O

InChI

1S/C4H9NO3/c1-2(6)3(5)4(7)8/h2-3,6H,5H2,1H3,(H,7,8)/t2-,3-/m0/s1

InChI key

AYFVYJQAPQTCCC-HRFVKAFMSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

E Meirzadeh et al.
Nature communications, 7, 13351-13351 (2016-11-09)
Doping is a primary tool for the modification of the properties of materials. Occlusion of guest molecules in crystals generally reduces their symmetry by the creation of polar domains, which engender polarization and pyroelectricity in the doped crystals. Here we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service