Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

208906

Sigma-Aldrich

Nitro Red

Dye content 50 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10N4Na2O9S2
CAS Number:
Molecular Weight:
512.38
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 50%

λmax

546 nm

SMILES string

[Na+].[Na+].Nc1c(\N=N/c2ccc(cc2)[N+]([O-])=O)c(cc3cc(cc(O)c13)S([O-])(=O)=O)S([O-])(=O)=O

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service