Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20591

Sigma-Aldrich

Butylsilane

≥97.0% (GC)

Synonym(s):

n-Butylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H12Si
CAS Number:
Molecular Weight:
88.22
Beilstein:
1730905
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

refractive index

n20/D 1.392

bp

56.7 °C (lit.)

density

0.68 g/mL at 20 °C (lit.)

SMILES string

CCCC[SiH3]

InChI

1S/C4H12Si/c1-2-3-4-5/h2-4H2,1,5H3

InChI key

YXMVRBZGTJFMLH-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

21.2 °F - closed cup

Flash Point(C)

-6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kyo Koizumi et al.
The journal of physiological sciences : JPS, 69(1), 65-77 (2018-05-16)
To investigate how the functional architecture is organized in layer 5 (L5) of the somatosensory cortex of a mouse in vivo, the input-output relationship was investigated using an all-optical approach. The neural activity in L5 was optically recorded using a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service