Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

205265

Sigma-Aldrich

2,2-Dimethylglutaric acid

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2CH2C(CH3)2COOH
CAS Number:
Molecular Weight:
160.17
Beilstein:
1768501
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

powder

mp

82-86 °C (lit.)

functional group

carboxylic acid

SMILES string

CC(C)(CCC(O)=O)C(O)=O

InChI

1S/C7H12O4/c1-7(2,6(10)11)4-3-5(8)9/h3-4H2,1-2H3,(H,8,9)(H,10,11)

InChI key

BTUDGPVTCYNYLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The vapour pressures of 2,2-dmethylglutaric acid at different temperatures was studied using Knudsen mass-loss effusion technique[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermodynamic study on the sublimation of succinic acid and of methyl-and dimethyl-substituted succinic and glutaric acids.
The Journal of Chemical Thermodynamics, 33(1), 23-31 (2001)
Ayesha Hafeez et al.
JPMA. The Journal of the Pakistan Medical Association, 70(1), 53-57 (2020-01-19)
To determine the pattern of various inherited metabolic disorders specifically through plasma amino acid and urine organic acid testing in high-risk paediatric population.. The cross-sectional retrospective study was conducted at the Armed Forces Institute of Pathology, Rawalpindi, Pakistan, and comprised
Hajime Nakatani et al.
Biotechnology and bioengineering, 116(2), 239-249 (2018-11-07)
In the cell surface display system, the distance of a surface-displayed molecule from the cell surface should influence its functionality due to the interference by other surface structures. For the purpose of developing this distance-variable surface display system, we utilized
M Birringer et al.
British journal of cancer, 88(12), 1948-1955 (2003-06-12)
Recent results show that alpha-tocopheryl succinate (alpha-TOS) is a proapoptotic agent with antineoplastic activity. As modifications of the vitamin E (VE) molecule may affect its apoptogenic activity, we tested a number of newly synthesised VE analogues using malignant cell lines.
Kyaw Aung et al.
The Plant cell, 32(3), 595-611 (2020-01-01)
A hallmark of multicellular organisms is their ability to maintain physiological homeostasis by communicating among cells, tissues, and organs. In plants, intercellular communication is largely dependent on plasmodesmata (PD), which are membrane-lined channels connecting adjacent plant cells. Upon immune stimulation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service