Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

201200

Sigma-Aldrich

5-Chloro-4-nitro-o-toluidine

40%

Synonym(s):

2-Amino-4-chloro-5-nitrotoluene, 5-Chloro-2-methyl-4-nitroaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H2(NO2)(CH3)NH2
CAS Number:
Molecular Weight:
186.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

contains

~60% water

concentration

40%

mp

164-167 °C (lit.)

functional group

chloro
nitro

SMILES string

Cc1cc(c(Cl)cc1N)[N+]([O-])=O

InChI

1S/C7H7ClN2O2/c1-4-2-7(10(11)12)5(8)3-6(4)9/h2-3H,9H2,1H3

InChI key

OKOSGBYZOWWAPH-UHFFFAOYSA-N

Application

5-Chloro-4-nitro-o-toluidine was used in the synthesis of 2-substituted 3,7,8-trichlorodibenzo-p-dioxins[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Romkes et al.
Cancer research, 47(19), 5108-5111 (1987-10-01)
The competitive receptor binding affinities of thirteen 2-substituted 3,7,8-trichlorodibenzo-p-dioxins to hepatic cytosol from rat, mouse, guinea pig, and hamster were determined with [3H]-2,3,7,8-tetrachlorodibenzo-p-dioxin as the radioligand. Multiple parameter linear regression analysis of the binding data from the four species gave

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service