Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

201162

Sigma-Aldrich

Lithium benzoate

99%

Synonym(s):

Benzoic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOLi
CAS Number:
Molecular Weight:
128.05
Beilstein:
3598089
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99%
form:
powder

Assay

99%

form

powder

mp

>300 °C (lit.)

SMILES string

[Li+].[O-]C(=O)c1ccccc1

InChI

1S/C7H6O2.Li/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);/q;+1/p-1

InChI key

LDJNSLOKTFFLSL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • Electrically conductive ultrananocrystalline diamond-coated natural graphite-copper anode for new long life lithium-ion battery: This study mentions lithium benzoate films used as coatings to improve the performance of NG-based anodes in lithium-ion batteries (Cheng et al., 2014).
  • Effect of permittivity on the ionic boundary layer upon protonation of lithium niobate: Discusses lithium benzoate′s role in the recombination process with lithium niobate crystals and its impact on the ionic boundary layer (Demin et al., 2023).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service