Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

200522

Sigma-Aldrich

Poly(ethylene-co-propylene-co-5-methylene-2-norbornene)

ethylene 70 wt. %

Synonym(s):

Ethylene-5-ethylidenenorbornene-propene copolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

composition

ethylene, 70 wt. %

viscosity

80, Mooney(lit.)

density

0.86 g/mL at 25 °C (lit.)

InChI

1S/C9H12.C3H6.C2H4/c1-2-8-5-7-3-4-9(8)6-7;1-3-2;1-2/h2-4,7,9H,5-6H2,1H3;3H,1H2,2H3;1-2H2/b8-2+;;

InChI key

MPXNNMASLYQCAH-SZPWSVBHSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chelsea L Dieck et al.
Cancer cell, 34(1), 136-147 (2018-07-11)
Activating mutations in the cytosolic 5'-nucleotidase II gene NT5C2 drive resistance to 6-mercaptopurine in acute lymphoblastic leukemia. Here we demonstrate that constitutively active NT5C2 mutations K359Q and L375F reconfigure the catalytic center for substrate access and catalysis in the absence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service