Skip to Content
MilliporeSigma
All Photos(1)

Documents

19999

Sigma-Aldrich

tert-Butyl hydroperoxide solution

packed in FEP bottles, ~5.5 M in nonane (over molecular sieve 4 Å)

Synonym(s):

1,1-Dimethylethyl hydroperoxide, 2-Hydroperoxy-2-methylpropane, TBHP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COOH
CAS Number:
Molecular Weight:
90.12
Beilstein:
1098280
MDL number:
UNSPSC Code:
12352120
PubChem Substance ID:
NACRES:
NA.22

form

liquid

quality

packed in FEP bottles

reaction suitability

reagent type: oxidant

concentration

~5.5 M in nonane (over molecular sieve 4 Å)

refractive index

n20/D 1.400

density

0.82 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

CC(C)(C)OO

InChI

1S/C4H10O2/c1-4(2,3)6-5/h5H,1-3H3

InChI key

CIHOLLKRGTVIJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Safe and stable oxidant for the Sharpless epoxidation

Reactant also used as a redox initiator and oxidant

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Org. Perox. F - Skin Corr. 1C - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X. Zhu, et al.,
Huaxue Xuebao, 69, 1107-1114 (2011)
K. Alagiri and K. R. Prabhu,
Tetrahedron, 67, 8544-8551 (2011)
B. Chaband, K.B. Sharpless
The Journal of Organic Chemistry, 44, 4202-4202 (1979)
Ramya Sivangala Thandi et al.
Nature communications, 11(1), 3535-3535 (2020-07-17)
Macrophages are professional phagocytes known to play a vital role in controlling Mycobacterium tuberculosis (Mtb) infection and disease progression. Here we compare Mtb growth in mouse alveolar (AMs), peritoneal (PMs), and liver (Kupffer cells; KCs) macrophages and in bone marrow-derived monocytes (BDMs).
Qicai Xue et al.
Chemical communications (Cambridge, England), 49(35), 3700-3702 (2013-03-29)
A new synthetic approach toward direct C-N bond formation through sp(3) C-H activation has been developed under metal-free conditions. Both primary and secondary benzylic C-H substrates could react smoothly with various amines to give only mono-amination products with good to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service