Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

199427

Sigma-Aldrich

1,5-Dimethyl-2-pyrrolecarbonitrile

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N2
CAS Number:
Molecular Weight:
120.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

54-56 °C (lit.)

functional group

nitrile

SMILES string

Cc1ccc(C#N)n1C

InChI

1S/C7H8N2/c1-6-3-4-7(5-8)9(6)2/h3-4H,1-2H3

InChI key

DRXOPQFEWDRGKT-UHFFFAOYSA-N

General description

The standard molar enthalpy of formation of 1,5-dimethyl-2-pyrrolecarbonitrile in the gaseous phase was studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Santos AFL and Ribeiro da Silva MA.
The Journal of Chemical Thermodynamics, 48, 194-200 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service