Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

199370

Sigma-Aldrich

1-(4-Nitrophenyl)-2-thiourea

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4NHCSNH2
CAS Number:
Molecular Weight:
197.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

206 °C (dec.) (lit.)

functional group

amine
nitro
thiourea

SMILES string

NC(=S)Nc1ccc(cc1)[N+]([O-])=O

InChI

1S/C7H7N3O2S/c8-7(13)9-5-1-3-6(4-2-5)10(11)12/h1-4H,(H3,8,9,13)

InChI key

BLYAANPIHFKKMQ-UHFFFAOYSA-N

Application

1-(4-Nitrophenyl)-2-thiourea was used as anion sensor in organic solvent.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chomchai Suksai et al.
Chemical Society reviews, 32(4), 192-202 (2003-07-24)
Chromogenic sensors for anions generally consist of two parts: anion receptors and chromophores. In this review, 6 types of chromogenic anion sensors are described, namely, NH-based hydrogen bonding, Lewis acid, metal-ion template, transition metal complexes, chromogenic guest displacement and chromoreactands.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service