Skip to Content
MilliporeSigma
All Photos(1)

Documents

198862

Sigma-Aldrich

3-Methyl-2-phenylpyridine

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N
CAS Number:
Molecular Weight:
169.22
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

≥97%

refractive index

n20/D 1.6030 (lit.)

density

1.065 g/mL at 25 °C (lit.)

SMILES string

Cc1cccnc1-c2ccccc2

Looking for similar products? Visit Product Comparison Guide

Application

Intermediate in the synthesis of 4-azafluorene and δ-carboline.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 14, 13-13 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service