Skip to Content
MilliporeSigma
All Photos(1)

Documents

196932

Sigma-Aldrich

3-Methoxy-5-(trifluoromethyl)aniline

99%

Synonym(s):

5-Methoxy-α,α,α-trifluoro-m-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(CF3)NH2
CAS Number:
Molecular Weight:
191.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

solid

mp

50-52 °C (lit.)

SMILES string

COc1cc(N)cc(c1)C(F)(F)F

InChI

1S/C8H8F3NO/c1-13-7-3-5(8(9,10)11)2-6(12)4-7/h2-4H,12H2,1H3

InChI key

VTFGJEYZCUWSAM-UHFFFAOYSA-N

Application

3-Methoxy-5-(trifluoromethyl)aniline was used in the synthesis of spleen tyrosine kinase inhibitors based on the phenylamino pyrimidine thiazole.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luc J Farmer et al.
Bioorganic & medicinal chemistry letters, 18(23), 6231-6235 (2008-10-22)
A series of SYK inhibitors based on the phenylamino pyrimidine thiazole lead 4 were prepared and evaluated for biological activity. Lead optimization provided compounds with nanomolar K(i)'s against SYK and potent inhibition in mast cell degranulation assays.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service