Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

196576

Sigma-Aldrich

4-Nitro-2-(trifluoromethyl)aniline

98%

Synonym(s):

2-Amino-5-nitrobenzotrifluoride, 4-Nitro-α,α,α-trifluoro-o-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3(CF3)NH2
CAS Number:
Molecular Weight:
206.12
Beilstein:
2121347
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

90-92 °C (lit.)

functional group

fluoro
nitro

SMILES string

Nc1ccc(cc1C(F)(F)F)[N+]([O-])=O

InChI

1S/C7H5F3N2O2/c8-7(9,10)5-3-4(12(13)14)1-2-6(5)11/h1-3H,11H2

InChI key

HOTZLWVITTVZGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Nitro-2-(trifluoromethyl)aniline undergoes diazotization and coupling with:
  • N-hydroxyalkylamino-5-napthols
  • pyrazolones and naptholsulphonic acids
  • N-(2-cyanoethyl)-N-(hydroxyalkyl)anilines

Application

4-Nitro-2-(trifluoromethyl)aniline was used in the synthesis of monoazo dyes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of fluorine substitution on color and fastness of monoazo dyes.
Dickey JB, et al.
Ind. and Eng. Chem., 45(8), 1730-1734 (1953)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service