Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

196339

Sigma-Aldrich

Methyl 2-oxo-1-pyrrolidineacetate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11NO3
CAS Number:
Molecular Weight:
157.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

refractive index

n20/D 1.472 (lit.)

density

1.131 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CN1CCCC1=O

InChI

1S/C7H11NO3/c1-11-7(10)5-8-4-2-3-6(8)9/h2-5H2,1H3

InChI key

HPDZYDTXCOFUOY-UHFFFAOYSA-N

Application

Methyl 2-oxo-1-pyrrolidineacetate may be used in the synthesis of N-[(disubstituted-amino)alkyl]-2-oxo-1-pyrrolidineacetamide derivatives.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D E Butler et al.
Journal of medicinal chemistry, 27(5), 684-691 (1984-05-01)
A series of N-[(dialkylamino)alkyl]-2-oxo-1- pyrrolidineacetamides was synthesized. The title compounds reversed electroconvulsive shock (ECS) induced amnesia in mice when administered subsequent to the ECS treatment and were inactive in a general observational test for central nervous system (CNS) activity. Active

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service