Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

196037

Sigma-Aldrich

4-Isopropylbenzyl alcohol

97%

Synonym(s):

Cumic alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHC6H4CH2OH
CAS Number:
Molecular Weight:
150.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.519 (lit.)

bp

135-136 °C/26 mmHg (lit.)

solubility

alcohol: miscible
diethyl ether: miscible
water: insoluble

density

0.982 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC(C)c1ccc(CO)cc1

InChI

1S/C10H14O/c1-8(2)10-5-3-9(7-11)4-6-10/h3-6,8,11H,7H2,1-2H3

InChI key

OIGWAXDAPKFNCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Isopropylbenzyl alcohol has mosquito repellent properties and has been isolated from Eucalyptus camaldulensis[1]. Oxidation of 4-isopropylbenzyl alcohol over TiO2-supported nano gold catalyst has been investigated[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective oxidation of alcohols to aldehydes and ketones over TiO2-supported gold nanoparticles in supercritical carbon dioxide with molecular oxygen.
Wang X, et al.
Applied Catalysis A: General, 349(1), 86-90 (2008)
New mosquito repellent from Eucalyptus camaldulensis.
Watanabe K, et al.
Journal of Agricultural and Food Chemistry, 41(11), 2164-2166 (1993)
Annemieke Strijkstra et al.
Applied and environmental microbiology, 80(24), 7592-7603 (2014-09-28)
The betaproteobacteria "Aromatoleum aromaticum" pCyN1 and "Thauera" sp. strain pCyN2 anaerobically degrade the plant-derived aromatic hydrocarbon p-cymene (4-isopropyltoluene) under nitrate-reducing conditions. Metabolite analysis of p-cymene-adapted "A. aromaticum" pCyN1 cells demonstrated the specific formation of 4-isopropylbenzyl alcohol and 4-isopropylbenzaldehyde, whereas with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service