Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

194956

Sigma-Aldrich

α-Methyl-α-propylsuccinimide

≥99%

Synonym(s):

3-Methyl-3-propylpyrrolidine-2,5-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13NO2
CAS Number:
Molecular Weight:
155.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥99%

mp

76-77 °C (lit.)

SMILES string

CCCC1(C)CC(=O)NC1=O

InChI

1S/C8H13NO2/c1-3-4-8(2)5-6(10)9-7(8)11/h3-5H2,1-2H3,(H,9,10,11)

InChI key

VXXGMHKGORIRTK-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samantha Y Chong et al.
Chemical communications (Cambridge, England), (39)(39), 4078-4080 (2006-10-07)
The principles of social and biological evolution have been combined in a Cultural Differential Evolution hybrid global optimization technique and applied to crystal structure solution.
M Riedmann et al.
Therapeutic drug monitoring, 3(4), 397-413 (1981-01-01)
Specific antiepileptic drug (AED) concentrations in serum are believed to cause therapeutic effects in individual patients. Communication between the clinician and the laboratory performing the drug level determinations is therefore essential for safe and effective medication. The now common practice

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service