Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

194166

Sigma-Aldrich

Methanol-d4

"100%", ≥99.96 atom % D, ≥99% (CP)

Synonym(s):

Methyl-d3 alcohol d, Tetradeuteromethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3OD
CAS Number:
Molecular Weight:
36.07
Beilstein:
1733278
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
To inquire about this product 194166, please contact your local MilliporeSigma office or dealer. Contact Technical Service

isotopic purity

≥99.96 atom % D

Assay

≥99% (CP)

form

liquid

expl. lim.

5.5-36.5 % (lit.)

technique(s)

NMR: suitable

impurities

≤0.250% water
water

refractive index

n20/D 1.326 (lit.)

bp

65.4 °C (lit.)

mp

-99 °C (lit.)

density

0.888 g/mL at 25 °C (lit.)

SMILES string

[2H]OC([2H])([2H])[2H]

InChI

1S/CH4O/c1-2/h2H,1H3/i1D3,2D

InChI key

OKKJLVBELUTLKV-MZCSYVLQSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Antithrombotic Phenolics from the Stems of Parthenocissus tricuspidata Possess Anti-inflammatory Effect.
Nguyen PH, et al.
Bull. Korean Chem. Soc., 35(6), 1763-1763 (2014)
Haoyue Zhang et al.
Clinica chimica acta; international journal of clinical chemistry, 508, 179-184 (2020-05-23)
To validate a liquid chromatography-tandem mass spectrometry (UPLC-MS/MS) method for the measurement of glycosaminoglycans (GAGs) in plasma and serum. To establish plasma, cerebrospinal fluid (CSF) and urine reference intervals. To compare GAGs in serum with that in urine and CSF
The Rates of Methoxyl Exchange of Camphor and Norcamphor Dimethyl Ketals in Methanol-d4.
Traylor TG and Perrin CL.
Journal of the American Chemical Society, 88(21), 4934-4942 (1966)
Cheng-Kun Lin et al.
Organic & biomolecular chemistry, 13(7), 2100-2107 (2014-12-20)
A straightforward synthesis of novel, 2-heterocyclyl polyhydroxylated pyrrolidines is described. Stereocontrolled additions of nucleophiles to cyclic nitrones generated the corresponding 2,3-trans adducts, allowing the synthesis of the corresponding pyrrolidines via key intermediates bearing an alkyne and a nitrile oxide. Three
Jon Kapla et al.
Physical chemistry chemical physics : PCCP, 17(34), 22438-22447 (2015-08-08)
The disaccharide trehalose (TRH) strongly affects the physical properties of lipid bilayers. We investigate interactions between lipid membranes formed by 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC) and TRH using NMR spectroscopy and molecular dynamics (MD) computer simulations. We compare dipolar couplings derived from DMPC/TRH

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service