Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

193704

Sigma-Aldrich

2,4,5-Trimethyloxazole

95%

Synonym(s):

Trimethyloxazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NO
CAS Number:
Molecular Weight:
111.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.442 (lit.)

bp

133-134 °C/760 mmHg (lit.)

density

0.957 g/mL at 25 °C (lit.)

SMILES string

Cc1nc(C)c(C)o1

InChI

1S/C6H9NO/c1-4-5(2)8-6(3)7-4/h1-3H3

InChI key

ZRLDBDZSLLGDOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4,5-Trimethyloxazole undergoes regio- and diastereoselective photocycloaddition reaction with aliphatic and aromatic aldehydes to yield erythro α-amino, β-hydroxy methyl ketones[1].

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxazole-Carbonyl photocycloadditions: selectivity pattern and synthetic route to erythro a-amino, ?-hydroxy ketones
Griesbeck AG, et al.
Chemical Communications (Cambridge, England), 589-590 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service