Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

192937

Sigma-Aldrich

exo-2-Norbornyl formate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O2
CAS Number:
Molecular Weight:
140.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.462 (lit.)

bp

65-67 °C/16 mmHg (lit.)

density

1.048 g/mL at 25 °C (lit.)

SMILES string

O=CO[C@H]1C[C@@H]2CC[C@H]1C2

InChI

1S/C8H12O2/c9-5-10-8-4-6-1-2-7(8)3-6/h5-8H,1-4H2/t6-,7+,8+/m1/s1

InChI key

SGXIEZNAOCVSKO-CSMHCCOUSA-N

General description

exo-2-norbornyl formate participates in ruthenium-catalyzed novel transformations of alkyl formates.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruthenium complex-catalyzed novel transformation of alkyl formates.
Kondo T, et al.
Journal of Organometallic Chemistry, 489(1), 83-91 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service