Skip to Content
MilliporeSigma
All Photos(1)

Documents

192538

Sigma-Aldrich

5-Chloro-2-(trichloromethyl)benzimidazole

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4Cl4N2
CAS Number:
Molecular Weight:
269.94
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

223-224 °C (dec.) (lit.)

SMILES string

Clc1ccc2[nH]c(nc2c1)C(Cl)(Cl)Cl

InChI

1S/C8H4Cl4N2/c9-4-1-2-5-6(3-4)14-7(13-5)8(10,11)12/h1-3H,(H,13,14)

InChI key

SIZGSKQSWJIWFP-UHFFFAOYSA-N

General description

The effect of 5-chloro-2-trichloromethyl benzimidazole on binding affinity of human growth hormone (hGH) (Thr175 → Gly–hGH) to the extracellular domain of the hGH receptor (Trp104 → Gly–hGHbp) has been investigated.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z Guo et al.
Science (New York, N.Y.), 288(5473), 2042-2045 (2000-06-17)
Mutations introduced into human growth hormone (hGH) (Thr175 --> Gly-hGH) and the extracellular domain of the hGH receptor (Trp104 --> Gly-hGHbp) created a cavity at the protein-protein interface that resulted in binding affinity being reduced by a factor of 10(6).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service