Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19162

Sigma-Aldrich

(R)-3-Butene-1,2-diol-1-(p-toluenesulfonate)

≥99.0% (HPLC)

Synonym(s):

(R)-1-Tosyloxy-3-buten-2-ol, (R)-2-Hydroxy-3-butenyl tosylate, (R)-3-Butene-1,2-diol 1-tosylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14O4S
CAS Number:
Molecular Weight:
242.29
Beilstein:
5907827
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

Assay

≥99.0% (HPLC)

optical activity

[α]20/D +7.7±0.5°, c = 1% in methanol

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1)S(=O)(=O)OC[C@H](O)C=C

InChI

1S/C11H14O4S/c1-3-10(12)8-15-16(13,14)11-6-4-9(2)5-7-11/h3-7,10,12H,1,8H2,2H3/t10-/m1/s1

InChI key

YQSCDBZHHLIPOI-SNVBAGLBSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service