Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

191515

Sigma-Aldrich

Methyl 4-oxo-3-piperidinecarboxylate hydrochloride

97%

Synonym(s):

3-Methoxycarbonyl-4-piperidone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11NO3 · HCl
CAS Number:
Molecular Weight:
193.63
Beilstein:
4273241
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
To inquire about this product 191515, please contact your local MilliporeSigma office or dealer. Contact Technical Service

Assay

97%

mp

~175 °C (dec.) (lit.)

SMILES string

Cl.COC(=O)C1CNCCC1=O

InChI

1S/C7H11NO3.ClH/c1-11-7(10)5-4-8-3-2-6(5)9;/h5,8H,2-4H2,1H3;1H

InChI key

NMAACQILAGCQPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl 4-oxo-3-piperidinecarboxylate hydrochloride has been used as starting reagent in the synthesis of:
  • nakadomarin A[1]
  • tert-butyl, picolyl and fluorinated analogs of capromorelin[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic approach towards nakadomarin A: efficient synthesis of the central tetracyclic core.
Nagata T, et al.
Tetrahedron Letters, 42(47), 8345-8349 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service