Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

190586

Sigma-Aldrich

4,4-Dimethyl-2-pentanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH2CH(OH)CH3
CAS Number:
Molecular Weight:
116.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.418 (lit.)

bp

137-137.5 °C/736 mmHg (lit.)

density

0.815 g/mL at 25 °C (lit.)

SMILES string

CC(O)CC(C)(C)C

InChI

1S/C7H16O/c1-6(8)5-7(2,3)4/h6,8H,5H2,1-4H3

InChI key

OIBKGNPMOMMSSI-UHFFFAOYSA-N

General description

Chemical ionization mass spectra of 4,4-dimethyl-2-pentanol has been studied[1].

Application

4,4-Dimethyl-2-pentanol has been used in the synthesis of tertiary 2-aminoquinoline derivatives[2].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew J Souers et al.
Bioorganic & medicinal chemistry letters, 14(19), 4873-4877 (2004-09-03)
A high-throughput screen was performed in order to identify chemotypes that are bound by the melanin concentrating hormone receptor-1 (MCHr1). A novel 2-amino-8-alkoxyquinoline compound (1) was identified and subsequently optimized using a parallel and automated procedure for the rapid production
Chemical ionization mass spectrometry. XII. Alcohols.
Field FH.
Journal of the American Chemical Society, 92(9), 2672-2676 (1970)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service