Skip to Content
MilliporeSigma
All Photos(1)

Documents

190551

Sigma-Aldrich

5-Diethylamino-2-pentanol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H21NO
CAS Number:
Molecular Weight:
159.27
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

97%

SMILES string

CCN(CC)CCCC(C)O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service