Skip to Content
MilliporeSigma
All Photos(1)

Documents

190136

Sigma-Aldrich

α-Ethylphenethyl alcohol

97%

Synonym(s):

1-Phenyl-2-butanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(C2H5)OH
CAS Number:
Molecular Weight:
150.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.516 (lit.)

bp

124-127 °C/25 mmHg (lit.)

density

0.989 g/mL at 25 °C (lit.)

SMILES string

CCC(O)Cc1ccccc1

InChI

1S/C10H14O/c1-2-10(11)8-9-6-4-3-5-7-9/h3-7,10-11H,2,8H2,1H3

InChI key

LYUPJHVGLFETDG-UHFFFAOYSA-N

Application

α-Ethylphenethyl alcohol may be used in chemical synthesis.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gregory R Tintinger et al.
Frontiers in immunology, 11, 621148-621148 (2021-03-16)
Although bedaquiline has advanced the treatment of multidrug-resistant tuberculosis (TB), concerns remain about the cardiotoxic potential of this agent, albeit by unexplored mechanisms. Accordingly, we have investigated augmentation of the reactivity of human platelets in vitro as a potential mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service