Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

189243

Sigma-Aldrich

4-Fluoro-2-methylaniline

96%

Synonym(s):

2-Amino-5-fluorotoluene, 4-Fluoro-o-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(CH3)NH2
CAS Number:
Molecular Weight:
125.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.537 (lit.)

bp

90-92 °C/16 mmHg (lit.)

density

1.126 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Cc1cc(F)ccc1N

InChI

1S/C7H8FN/c1-5-4-6(8)2-3-7(5)9/h2-4H,9H2,1H3

InChI key

KMHLGVTVACLEJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Fluoro-2-methylaniline was used in the synthesis of 4-fluoro-N-[(E)-{7-methoxy-2-[4-(methylsulfanyl)phenyl]-1-benzofuran-5-yl}methylidene]-2-methylaniline[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SYNTHESIS OF SCHIFF BASES OF 7-METHOXY-2-[4-(METHYLSULFANYL) PHENYL]-1-BENZOFURAN-5-CARBOXALDEHYDE AND ANIMES AND HYDRAZIDE.
Thorat BR, et al.
Heterocyclic Letters, 4(1), 41-53 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service