Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

189049

Sigma-Aldrich

2-Benzyloxyaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H4NH2
CAS Number:
Molecular Weight:
199.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

35-39 °C (lit.)

SMILES string

Nc1ccccc1OCc2ccccc2

InChI

1S/C13H13NO/c14-12-8-4-5-9-13(12)15-10-11-6-2-1-3-7-11/h1-9H,10,14H2

InChI key

PLPVLSBYYOWFKM-UHFFFAOYSA-N

General description

2-Benzyloxyaniline is an important intermediate in organic synthesis.

Application

2-Benzyloxyaniline was used to investigate crystal structure of benzyloxybenzoic acids and benzyloxyanilines by X-ray powder diffraction technique. It was also used in the synthesis of 2-benzyloxyphenylhydrazine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural study of four benzyloxybenzoic acids and benzyloxyanilines using X-ray powder diffraction: interplay of strong and weak intermolecular interactions.
Chattopadhyay B, et al.
CrystEngComm, 15(6), 1077-1085 (2013)
Selective alkylation of aminophenols.
Wang R and Xu J.
ARKIVOC (Gainesville, FL, United States), 293-299 (2010)
Catherine Roullier et al.
Bioorganic & medicinal chemistry letters, 20(15), 4582-4586 (2010-06-24)
A new aryl-hydrazide l-glutamic acid derivative, pygmeine (3), was isolated from a methanolic extract of Lichina pygmaea, a marine lichen. Synthetic derivatives obtained via a two-step coupling of l-glutamic acid with phenylhydrazine moieties were useful to elucidate the structure of
H Ottosson et al.
Scientific reports, 6, 36692-36692 (2016-11-09)
A new concept for treatment of infections is induction of our own antimicrobial peptides and the presented novel class of inducer, aroylated phenylenediamines (APDs), gives up to 20 to 30-fold induction of the human antimicrobial peptide LL-37, in vitro. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service