Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

188972

Sigma-Aldrich

3,3-Diphenyl-1-propanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2CHCH2CH2OH
CAS Number:
Molecular Weight:
212.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.584 (lit.)

bp

185 °C/10 mmHg (lit.)

density

1.067 g/mL at 25 °C (lit.)

SMILES string

OCCC(c1ccccc1)c2ccccc2

InChI

1S/C15H16O/c16-12-11-15(13-7-3-1-4-8-13)14-9-5-2-6-10-14/h1-10,15-16H,11-12H2

InChI key

IDCXQMVSIIJUEH-UHFFFAOYSA-N

General description

3,3-Diphenyl-1-propanol undergoes cobalt-catalyzed hydroformylation reaction to give the corresponding aldehyde and 1,1-diphenylethylene.

Application

3,3-Diphenyl-1-propanol has been used in the preparation of novel and very potent prostacyclin (PGI2) agonists.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular design of novel PGI2 agonists without PG skeleton. IV.
Hamanaka N, et al.
Bioorganic & Medicinal Chemistry Letters, 5(10), 1083-1086 (1995)
Is the hydroformylation reaction reversible?
Kim C, et al.
Journal of Organometallic Chemistry, 279(1), 159-164 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service