Skip to Content
MilliporeSigma
All Photos(1)

Documents

188794

Sigma-Aldrich

3,4-Dimethylbenzyl alcohol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3CH2OH
CAS Number:
Molecular Weight:
136.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

218-221 °C (lit.)

mp

62-65 °C (lit.)

SMILES string

Cc1ccc(CO)cc1C

InChI

1S/C9H12O/c1-7-3-4-9(6-10)5-8(7)2/h3-5,10H,6H2,1-2H3

InChI key

OKGZCXPDJKKZAP-UHFFFAOYSA-N

Application

3,4-Dimethylbenzyl alcohol was used in determination of toxicities of pseudocumene and its metabolites for E.coli JM101.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hye Won Kim et al.
Molecules (Basel, Switzerland), 24(7) (2019-04-17)
Styrene can be formed by the microbial metabolism of bacteria and fungi. In our previous study, styrene was determined as a spoilage marker of Fuji apples decayed by Penicillium expansum, which is responsible for postharvest diseases. In the present study
Bruno Bühler et al.
Applied and environmental microbiology, 68(2), 560-568 (2002-02-02)
Xylene monooxygenase of Pseudomonas putida mt-2 catalyzes multistep oxidations of one methyl group of toluene and xylenes. Recombinant Escherichia coli expressing the monooxygenase genes xylM and xylA catalyzes the oxygenation of toluene, pseudocumene, the corresponding alcohols, and the corresponding aldehydes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service